Potassium Hydrogen Phthalate

Supplier: ALDON CORP SE

PP0566-100GR PP0566-500GR PP0567-500ML
470302-044EA 30.75 CAD
470302-044 470302-046 470225-706
Potassium Hydrogen Phthalate
Chemicals for Science Education

CAS Number: 877-24-7
Formula: C8H5KO4
Density: 1.636 g/mL
Freezing Point: 295-300°C
Solubility: Water
Synonyms: Potassium Biphthalate, Potassium Acid Phthalate
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR