Potassium Ferrocyanide Trihydrate

Supplier: ALDON CORP SE

PP0550-100G PP0550-100GR
470302-122EA 36.25 CAD
470302-122 470302-126
Potassium Ferrocyanide Trihydrate
Chemicals for Science Education

CAS Number: 14459-95-1
Formula Weight: 422.41
Formula: K4Fe(CN)6·3H2O
Density (g/mL): 1.853
Freezing Point (°C): >70
Solubility: Water
Synonyms: Yellow Prussiate of Potash
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR