Potassium Ferricyanide

Supplier: ALDON CORP SE

PP0539-CONC PP0530-100G PP0530-500G PP0530-100GR
470302-118EA 19.2 CAD
470302-118 470302-114 470302-116 470302-120
Potassium Ferricyanide
Chemicals for Science Education

CAS Number: 13746-66-2
Formula Weight: 329.26
Formula: K3Fe(CN)6
Hazard Info: Oxidizer, Toxic
Density (g/mL): 1.85
Solubility: Water and Alcohol
Synonyms: Red Prussiate of Potash
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR