Potassium Dichromate

Supplier: ALDON CORP SE

PP0521-50ML PP0516-500G PP0516-500GR PP0516-100G
470302-104EA 20.3 CAD
470302-104 470302-110 470302-112 470302-108
Potassium Dichromate
Chemicals for Science Education

CAS Number: 7778-50-9
Formula Weight: 294.19
Formula: K2Cr2O7
Density (g/mL): 2.67
Freezing Point (°C): 398
Solubility: Water
Synonyms: Potassium Bichromate
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR