Potassium Chloride
Supplier: ALDON CORP SE
CAS Number: 7447-40-7
Formula: KCl
Density: 1.986—1.990 g/mL
Boiling and Freezing Point: 1500 °C, 772 to 776 °C
Solubility: Water and Alcohol
Synonyms: Muriate of Potash, Potassium Muriate, Potassium Monochloride
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...