Potassium Chlorate
Supplier: ALDON CORP SE
CAS Number: 3811-04-9
Formula: KClO3
Density: 2.34 g/mL
Boiling and Freezing Point: 400°C, 368°C
Solubility: Boiling Water
Synonyms: Chlorate of Potash, Chloric Acid Potassium Salt, Potassium Oxymuriate
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is an explosive hazard, oxidizer, and irritant.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...