Potassium Bromide

Supplier: ALDON CORP SE

PP0408-500ML PP0409-CONC PP0400-100G PP0400-500G PP0400-100GR PP0415-500ML
470302-056EA 23.45 CAD
470302-056 470302-058 470302-060 470302-062 470302-064 470225-736
Potassium Bromide
Chemicals for Science Education

CAS Number: 7758-02-3
Formula Weight: 119.01
Formula: KBr
Density (g/mL): 2.749
Boiling Point (°C): 1435
Freezing Point (°C): 760
Solubility: Water and Alcohol
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR