Potassium

Supplier: ALDON CORP SE

PP0635-10G PP0635-25G PP0635-100G PP0635-1G
470302-182EA 252.8 CAD
470302-182 470302-186 470302-184 470302-288
Potassium
Chemicals for Science Education

CAS Number: 7440-09-7
Formula: K
Density: 0.86 g/mL
Boiling and Freezing Point: 774°C, 63°C
Synonyms: Kalium
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable and explosive.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR