Porcelain Spot Plates
Supplier: SILICO & CHEMICO PORCELAIN SE
J-291 J-289
470039-540EA 159.5 CAD
470039-540 470153-506
Porcelain Spot Plates
Staining Tiles
These plates are for use in microchemistry experiments.
  • Glazed Porcelain
  • Acid-Resistant
  • Chemically-Inert
  • Plates Withstand up to 1000°F
The glazed porcelain spot plates are acid resistant and chemically inert.
They can withstand temperatures up to 1000°F.
Order Now
SPECIFICATIONS
Environmentally Preferable