Polypropylene Short Stem Funnels, 60°, Bel-Art
Supplier: Bel-Art
F14676-0075_SE F14676-0055 F14676-0065_SE 4250-0035 4250-0160
470145-358BG 96.4 CAD
470145-358 470145-362 470201-734 470211-836 470211-840
Polypropylene Short Stem Funnels, 60°, Bel-Art
Funnels Filtration Funnels Filter Funnels
The external ribbing on this funnel prevents air lock.
  • Funnel Has 60° Angle
  • Internal Ribs for Speedier Filtrations
  • External Ribbing Prevents Air Lock
  • Autoclavable
The funnel has a 60° angle. The internal ribs allow for speedier filtrations.
It is autoclavable.