Polypropylene Beakers

Supplier: Bel-Art
F26219-0000 F26214-0000
470145-340CS 586.8 CAD
470145-340 470145-342
Polypropylene Beakers
Beakers
Translucent Polypropylene Beakers.

  • Raised graduations ±5% accuracy
  • Stack without sticking together
  • Autoclavable up to 121°C

Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR