Polypropylene Beaker Sets
Supplier: Dynalon
522135-0001 522135-0003 522135-0001
470020-818KT 90.9 CAD
470020-818 470020-820 490012-214
Polypropylene Beaker Sets
Beakers
Unbreakable Polypropylene
  • Autoclavable
  • Chemical-resistant
These beaker sets are autoclavable up to 145°C and chemical resistant. All feature pour spouts and metric graduation scales.