Polymethylpentene High-Speed Funnels, Bel-Art
Supplier: Bel-Art
F14640-0000_SE F14642-0000_SE F14642-0000_SE F14646-0000_SE
470122-356CS 533.55 CAD
470122-356 470122-358 470145-058 470200-376 470200-378
Polymethylpentene High-Speed Funnels, Bel-Art
Funnels Filtration Funnels Filter Funnels
For Non-Vacuum, High-Speed Filtration
  • Internal ribs increase the filtration speed of filter paper
  • Vertical outer ribs prevent airlock
  • Unaffected by corrosive liquids
  • Autoclavable; resists heat up to 177 °C