Polymethylpentene Erlenmeyer Flask with Screw Cap
Supplier: Thermo Fisher Scientific
4109-1000 4109-0250 4109-0500
470203-530CS 682.3 CAD
470203-530 470211-824 470211-826
Polymethylpentene Erlenmeyer Flask with Screw Cap
Flasks Erlenmeyer Flasks
PMP flasks with secure leak-proof seal.
  • Translucent
  • Includes screw cap
  • Resistant to most acids, bases, alcohols, esters, and aldehydes
  • Autoclavable