Polyethylene Wash Bottles
Supplier: VWR International
470191-300 470191-302 470191-312
470191-300CS 238.8 CAD
470191-300 470191-302 470191-312
Polyethylene Wash Bottles
Bottles Wash Bottles
Unbreakable wash bottles for student use.
  • Polyethylene, narrow mouth construction
  • Closures included
Cap and delivery tube are one-piece construction to eliminate leakage. The inside diameter of the delivery tip is carefully drawn to give a fine stream of liquid.