Pocket Magnifiers
Supplier: Pinetree Instruments
23202-3 23203-3 23201-3
470021-804CS 1543.5 CAD
470021-804 470021-806 470021-802
Pocket Magnifiers
Magnifiers
These pocket magnifiers are economical enough to outfit an entire class.
  • Handy plastic magnifiers
  • Ideal for younger students
  • Folds conveniently into attached case
  • Double/triple lenses can be stacked to increase magnification
Handy plastic magnifiers, with your choice of single, double, or triple lenses, are ideal for younger students. Each folds conveniently into an attached oval case. The 10X and 15X models’ double and triple lenses can be used separately or “stacked” to increase the magnification.
Dimensions: 1 1/2L x 1 1/4W x 1/4"H folded, 2 3/4"L extended.