Plastic Spring Wave
Supplier: NADA SCIENTIFIC LTD.
C15-4103
470039-692EA 51.3 CAD
470039-692
Plastic Spring Wave
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Observe phase reversal at the fixed end of wave pulses with this spring used to test waves in fundamental and multiple vibrations.
  • Ø×L: 2×50 cm
  • Experiments by students
With many versatile applications, it can also be used in experiments to determine the speed of propagation of transverse and longitudinal waves. Lightweight, the non-tangling plastic spring is ideal for student use.