Plastic Knife Switches
Supplier: PRECISION MOULDED POLYMERS LIMITED
1350303210000 97-1104
470005-868EA 14.6 CAD
470005-868 470005-870 470020-556
Plastic Knife Switches
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Plastic Knife Switches are easy to connect.
  • Easy-To-Connect
  • Unbreakable
  • Teaching Tool for Open Circuit Lessons
  • Rated at 5A
  • Two Configurations Available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits.
These unbreakable switches are available in two configurations. They are rated at 5A.