Plastic Coated Clear Glass Safety Bottles
Supplier: DWK Life Sciences (KIMBLE)
220735 220736
470205-584CS 531.24 CAD
470205-584 470205-586
Plastic Coated Clear Glass Safety Bottles
Bottles Glass Bottles
Provides a high degree of safety.
  • Coating protects against breakage, slips, and handling of hazardous chemicals
  • Cap is resistant to organic solvents