Plastic Coated Amber Glass Safety Bottles
Supplier: DWK Life Sciences (KIMBLE)
220935 220936
470205-590CS 622.25 CAD
470205-590 470205-592
Plastic Coated Amber Glass Safety Bottles
Bottles Glass Bottles
Provides a high degree of safety.
  • Coating protects against breakage, slips, and handling of hazardous chemicals
  • Designed specifically for light-sensitive chemicals
  • Cap is resistant to organic solvents