Plastic Beads
Supplier: Avantor
470015-258PK 15.8 CAD
470015-258 470023-956 470023-958
Plastic Beads
Beads Plastic Beads
Packages of approximately 100.
  • 4 mm Red bead = Acrylic
  • 8 mm Navy bead = Acrylic
  • 12 mm White bead = Acetate
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable