Plant Light and Water Tester
Supplier: LUSTER LEAF PRODUCTS INC SE
1830-A
470175-074CS 348.7 CAD
470175-074
Plant Light and Water Tester
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
A slender probe gauges the soil mixture at the roots of a plant, while a light sensor on the front of the unit measures the amount of ambient light.
  • Size: 4L×2½W×1½"H
  • Probe length: 6"
The light-weight unit needs no batteries, so there is nothing to replace or wear out. It includes a booklet with information on light and water requirements for over 150 plants.