Plant Heating Mat
Supplier: HYDROFARM GARDEN CENTER SALES
MT10006
470021-112EA 68.4 CAD
470021-112
Plant Heating Mat
Gardening Supplies
Jump start your seeds with this heating mat that provides even, gentle warmth and encourages speedy germination for seeds and young plants.
The mat fits under common plant trays and warms the rooting area 10–20 degrees above the ambient temperature. The instructions for lighting, watering, and planting are printed on the mat.
20 W operation, 110 VAC. Size: 20"L x 10"W.