Plankton Dip Net
Supplier: Cumings
BW-20-X
470106-844EA 42.75 CAD
470106-844
Plankton Dip Net
Animal Supplies Animal Nets
Small Mesh Size to Collect More Specimens
  • Reinforced for More Durability
Collect microscopic specimens with this extra-fine mesh dip net. The net is re-inforced with a vinyl net saver, and a wooden 51 cm handle provides an excellent grip. Size: 19×15×7 cm.