Planck's Constant Determination Box
Supplier: VWR International
PH1365
470018-432EA 146.55 CAD
470018-432
Planck's Constant Determination Box
Educational Materials Physics Educational Materials Modern Physics Learning Activities
This box shows Planck's Constant in action.
  • For Determining Approximate Value of Planck's Constant
  • Seven LEDs from 470 nm to 940 nm
  • Users Can Plot a Graph of Energy Input
This apparatus is designed to determine the approximate value of Planck's Constant using the elbow voltage in the characteristic curves of a number of LEDs. There are seven LEDs mounted on the box with sockets for voltage and current measurements. By monitoring the voltage at which each LED begins to emit light, users can plot a graph of energy input as a function of light emitted frequency and deduce an approximate value of Planck's Constant.
An external power supply for 9V is required. A 9V battery with suitable connecting cords can also be used.