Ward's® Live Terrestrial Isopods (Pill Bugs and Sow Bugs)
Supplier: TONY JEFFRIES
0 875520 6703112 6703115 DEFAULT VPN
470176-302PK 83.65 CAD
470176-302 470180-310 470180-312 470189-262 470191-048 470191-112 470309-752 470313-206 470313-208
Ward's® Live Terrestrial Isopods (Pill Bugs and Sow Bugs)
Specimens Preserved Specimens Preserved Animal Materials
Often mistaken for beetles, these wingless crustaceans live their whole lives on land, feeding on decaying organic matter.
  • Easy to maintain in the laboratory or classroom
  • Do not bite or sting
  • Pill and Sow Bugs are crustaceans
  • Demonstrate differences in defensive behaviors between the two specimens
Pill Bugs (Armadillium vulgare) roll themselves into a ball if agitated.
Sow Bugs (Oniscus asellus) can be identified by their posterior "tail like" appendages, and will play dead when agitated. They are unable to roll into a ball.
Ordering information: These products contain living or perishable materials. Both Pill Bugs and Sow Bugs are easy to maintain in the laboratory or classroom. They prefer dark, damp locations such as under logs, leaves or rocks, and are most active at night. They are detritivores, and feed mostly on decaying organic matter.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.