Preserved Pig Kidney
Supplier: Avantor
WARD470001-664 WARD470001-662 WARD470001-666 WARD470001-754 WARD470001-670 WARD470001-668 WARD470001-660
470001-670PK 229 CAD
470001-670 470001-660 470001-664 470001-662 470001-668 470001-666 470001-754
Preserved Pig Kidney
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved pig kidneys. Available in vacuum packs.
  • High quality preservation
  • Mammalian organ dissection
  • Kidney dissection
Pig kidneys are available as plain (uninjected specimens) and well as double and triple injected to show additional detail. Double injected specimens highlight arteries and veins highlight in red and blue. Triple injected pig kidneys offer a third yellow injection to highlight the ureters.
Pig kidneys may contain a small cut as part of a USDA inspection procedure. This will not compromise specimen quality.