Kemtec® Photosynthesis Reaction Activities
Supplier: Kemtec
2-517 2-516
470006-236KT 425.25 CAD
470006-236 470007-840
Kemtec® Photosynthesis Reaction Activities
Educational Materials Biology Educational Materials Plant Learning Activities
Two activities to explore photosynthesis.
  • Examine photosynthesis in two parts
  • Examine one part or both
Photosynthesis I examines the hill reaction in isolated chloroplasts; splitting water to yield oxygen and release hydrogen. The proteolysis of water in the course of the reaction is measurable using a spectrophotometer or by visual examination due to the bleaching of the hydrogen acceptor. The hill reaction is run in the light and in the dark to demonstrate the light dependence of the reaction.
Photosynthesis II examines the phosphorylation ofMore Product Information