Phosphoric Acid

Supplier: ALDON CORP SE

PP0239-500MLR PP0239-2.5LR
470302-024EA 86.2 CAD
470302-024 470302-022
Phosphoric Acid
Chemicals for Science Education

CAS Number: 7664-38-2
Formula Weight: 98.00
Formula: H3PO4
Hazard Info: Corrosive, Toxic, Irritant
Density (g/mL): >1.573
Boiling Point (°C): 135-158
Freezing Point (°C): -17-21
Solubility: Water and Alcohol
Synonyms: ortho-Phosphoric Acid
Shelf Life (months): 36
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR