Ward's® Live Coliphage Suspensions
Supplier: Avantor
470176-272EA 28.45 CAD
470176-272 470179-028 470179-030 470179-032 470179-036 470179-038
Ward's® Live Coliphage Suspensions
Specimens Live Specimens Viruses - Living Material
A collection of some of the more popular forms of coliphage. The successful culture of coliphage requires growing in the appropriate host bacterium (available separately)
Liquid coliphage suspension are routinely tested for quality, purity, and titer. They contain DNA or RNA supplied as a filtered tryptic soy broth suspension of relative infective titer (10-7 plaque-forming units per mL). Several varieties of coliphage cultures are available.
Ordering information: Contents include: PhiX174: Minute polyhedral form, 25 ?m in dia. Circular single-stranded DNA close relative of S13 phage.More Product Information