Phenolphthalein

Supplier: ALDON CORP SE

PP0161-1L PP0161-100ML PP0161-500ML PP0160-1L PP0160-100ML PP0160-30ML PP0160-500ML PP0159-500ML PPO160-PK/2 PP0159-1L PP0140-100G PP0140-25G PP0140-100GR
470301-978EA 38.65 CAD
470301-978 470301-980 470301-982 470301-984 470301-986 470301-988 470301-992 470301-996 470301-990 470301-994 470301-998 470302-000 470302-002
Phenolphthalein
Chemicals for Science Education

CAS Number: 77-09-8
Formula: C20H14O4
Density: 1.299 g/mL
Solubility: Alcohol and Alkalis
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR