Adjustable Height Student Lab Tables, Phenolic Top
Supplier: Diversified Woodcrafts
P8104 P8204 P8304 P8604
470205-358EA 2533.35 CAD
470205-358 470205-364 470205-370 470205-376
Adjustable Height Student Lab Tables, Phenolic Top
Furniture Tables and Workbenches
These sturdy adjustable height tables are manufacuted from a 4.5" solid oak frame with exposed metal corner braces and powder coated legs with adjustable glides. Available in four different sizes.
  • Adjusting only one end creates an incline table for physics
  • 1" phenolic work surface
  • For use in elementary grades 27”, for display work raise to 39”
  • Adjusts in 0.75" (2 cm) increments
Load capacity: 500 lbs.
This table can be adjusted in less than 30 seconds without the use of tools.