Phenol Red

Supplier: ALDON CORP SE

PP0175-100ML PP0175-500ML PP0180-500ML PP0170-5GR PP0180-100ML
470301-964EA 11.4 CAD
470301-964 470301-966 470301-970 470301-974 470301-968
Phenol Red
Chemicals for Science Education

CAS Number: 34487-61-1
Formula: C19H13NaO5S
Solubility: Water
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is a toxic irritant.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR