Phenol

Supplier: ALDON CORP SE

PP0120-500ML PP0130-500ML
470301-962EA 190.05 CAD
470301-962 470301-976
Phenol
Chemicals for Science Education

CAS Number: 108-95-2
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Toxic, Irritant (absorption and inhalation)
Density (g/mL): 1.05
Boiling Point (°C): 181
Freezing Point (°C): 5-17
Solubility: Water and Organic Solvents
Shelf Life (months): 12
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR