Petri Dish Labels
Supplier: DIVERSIFIED BIOTECH
PSTK-1000 PSTK-2000
470202-494PK 69.15 CAD
470202-494 470202-496
Petri Dish Labels
Labels
Clever alternative to using colony counters.
  • Made with water resistance ink
  • Remain firmly attached in temperatures ranging from 42 to 4 °C
  • Simply remove protective packing and apply to petri dish
  • Can be stored in incubator or refrigerator adhering to the temperatures mentioned above
  • Also, they can be held comfortably in place for streaking or colony transfer
Simply attach this transparent, waterproof sticker to the bottom of your Petri dish. The equally-shaped divisions of the grid are ideal for colony counts, while the 6-section is perfect for sensitivity studies.
Packaging: Packages of 36.