Slippable Petri Dishes, 100 mm
Supplier: Avantor
25384-302 WARD470151-288
470151-288PK 6.8 CAD
470151-288 470210-568
Slippable Petri Dishes, 100 mm
Culture Dishes Petri Dishes
These slippable polystyrene Petri dishes provide optimal flatness.
  • Unsurpassed in optical calrity - Ideal for micro photography
  • Engineered for optimum flatness to provide uniform agar thickness
  • Manufactured from pure virgin polystyrene
Offered in a convenient pack of six for small batches.
Packaging: Pack of six.