Pepsin
Supplier: ALDON CORP SE
PP0070-100G
PP0070-25G
470301-942EA
70.55
CAD
470301-942
470301-944
Pepsin
Chemicals for Science Education
CAS Number: 9001-75-6
Freezing Point: 100 °C
Solubility: Water
Synonyms: Pepsinum
Shelf Life: 12 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...