Pentane
Supplier: ALDON CORP SE
CAS Number: 109-66-0
Formula Weight: 72.15
Formula: C5H12
Density (g/mL): 0.626
Boiling Point (°C): 36
Freezing Point (°C): -130
Synonyms: n-Pentane, Amyl Hydride
Shelf Life (months): 36
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...