Parafilm
Supplier: Heathrow Scientific
120265 HS234526A
470144-226CS 715.35 CAD
470144-226 470201-930
Parafilm
Seals General Purpose Sealing Films
Flexible, self-adhesive, waterproof plastic for closures, mounting and more.
Adheres quickly and firmly to itself, covering culture tube or flask mouths and sealing petri dish edges. Excellent resistance to air, other gases, alcohol, common acids, and moderately concentrated alkalies.
Order Now
SPECIFICATIONS
Environmentally Preferable