Paraffin Wax, Refined White

Supplier: ALDON CORP SE

PP0051-454G PP0056-500G
470045-564EA 29.05 CAD
470045-564 470301-926
Paraffin Wax, Refined White
Chemicals for Science Education

CAS Number: 64742-43-4
Density (g/mL): 0.8
Freezing Point (°C): 48.8-71
Solubility: Warm Alcohol, Chloroform, Petroleum Ether, Olive Oil, Organic Solvents
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR