Pancreatin

Supplier: ALDON CORP SE

PP0010-100G PP0010-2G PP0010-10G PP0010-25G
470301-552EA 76.85 CAD
470301-552 470301-916 470301-918 470301-922
Pancreatin
Chemicals for Science Education

CAS Number: 8049-47-6
Solubility: Partially in Water
Synonyms: Pancreatic Enzyme
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR