Mammalian Ovary
Supplier: Avantor
470176-970EA 11 CAD
470176-970 470177-012
Mammalian Ovary
Slides Prepared Slides
Ovary (Mammalian, genus can vary) oogenesis and mature follicles. Ovary (Cat) Graafian follicles. Mature follicles with germ hill, or cumulus oophorous, and large ovum.