Onchocerca volvulus, In Situ Slide
Supplier: Avantor
WARD470182-156
470182-156EA 25.69 CAD
470182-156
Onchocerca volvulus, In Situ Slide
Slides Prepared Slides
Species of Nematode
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Nodule showing adults packed with eggs and larvae. Causes onchocerciasis, fibrous skin tumors appearing as nodules. The organism may also be the agent of nodding syndrome.