Ward's® Oligoclase
Supplier: Avantor
470014-632PK 38.55 CAD
470014-632 470014-634 470025-714
Ward's® Oligoclase
Specimens Mineral Specimens Classroom Minerals
Feldspar group; white-to-gray cleavages of sodic plagioclase; some show twinning.
Ordering information: WARD’S offer different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes Several of the smaller pieces are suitable for classroom study, while the larger specimens in the one kilogram pack can be cut to sizes that meet your specific needs. The number and size of pieces will vary.