Octyl Alcohol

Supplier: ALDON CORP SE

OX0005-500ML OX0005-100ML
470301-904EA 117.55 CAD
470301-904 470301-902
Octyl Alcohol
Chemicals for Science Education

CAS Number: 111-87-5
Formula: CH3CH26CH2OH
Density: 1.48 g/mL
Boiling Point: 195°C
Solubility: Miscible with Alcohol, Chloroform, and Mineral Oil
Synonyms: 1-Octanol, Capryl Alcohol, Octan-1-ol
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This item is flammable.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR