Nylon Semimicro Centrifuge Tube Brush
Supplier: Mill-Rose
17030-006
470120-522EA 5.35 CAD
470120-522
Nylon Semimicro Centrifuge Tube Brush
Brushes
Total length: 18 cm; bristle section length: 7.6 cm; tapered dia: from 1.3 cm to 5 mm.