Nitric Acid
Supplier: ALDON CORP SE
CAS Number: 7697-37-2
Formula: HNO3
Density: 1.37—1.42 g/mL
Boiling and Freezing Point: 120 to 122 °C, -22 to -41°C
Solubility: Miscible with Water
Synonyms: Azotic Acid
Shelf Life: 12 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is a corrosive and toxic oxidizer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...