Nigrosin
Supplier: ALDON CORP SE
NN0210-100ML
NN0206-25G
470301-870EA
27.8
CAD
470301-870
470301-872
Nigrosin
Chemicals for Science Education
CAS Number: 8005-03-6
Solubility: Water
Synonyms: Acid Black 2
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...