Nickel Sulfate 6-Hydrate

Supplier: ALDON CORP SE

NN0165-100GR NN0165-500G NN0166-500ML
470301-862EA 38.1 CAD
470301-862 470301-864 470225-744
Nickel Sulfate 6-Hydrate
Chemicals for Science Education

CAS Number: 10101-97-0
Formula Weight: 262.86
Formula: NiSO4·6H2O
Hazard Info: Toxic
Density (g/mL): 2.07
Solubility: Water and Alcohol
Synonyms: Nickelous Sulfate 6-Hydrate
Shelf Life (months): 12
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR