Nickel Nitrate 6-Hydrate

Supplier: ALDON CORP SE

NN0150-100G NN0150-500G NN0161-500ML NN0159-CONC
470301-854EA 32.25 CAD
470301-854 470301-856 470301-858 470301-860
Nickel Nitrate 6-Hydrate
Chemicals for Science Education

CAS Number: 13478-00-7
Formula Weight: 290.81
Formula: Ni(NO3)2·6H2O
Hazard Info: Suspected Human Carcinogen, Toxic
Density (g/mL): 2.065
Boiling Point (°C): 138
Freezing Point (°C): 57
Solubility: Water, Alcohol, and Ammonium Hydroxide
Synonyms: Nickel Nitrate Hexahydrate, Nickelous Nitrate 6-Hydrate
Shelf Life (months): 12
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR